Learn More
Calcium chloride dihydrate, ACS, 99.0-105.0%, Thermo Scientific Chemicals
£43.92 - £96.08
Chemical Identifiers
CAS | 10035-04-8 |
---|---|
Molecular Formula | CaCl2·2H2O |
MDL Number | MFCD00149613 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11367608
|
Thermo Scientific Alfa Aesar
033296.36 |
500 g |
£43.92
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11377608
|
Thermo Scientific Alfa Aesar
033296.A3 |
2 kg |
£96.08
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium chloride dihydrate is widely used as an ingredient in ceramic slipware, cosmetic and food items. It acts as a pH buffer in swimming pool water. It provides inhibition of swelling clays in the water phase of invert emulsion drilling fluids. It is employed in the oil industry to increase the density of solids-free brines. It is used as a flux in the Davy process for the manufacture of sodium through the electrolysis of molten sodium chloride. It finds an important application as an additive in plastics, fire extinguishers, blast furnaces and in wastewater treatment as a drainage aid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10035-04-8 | |
CaCl2·2H2O | |
MFCD00149613 |
Specifications
10035-04-8 | |
99.0-105.0% | |
MFCD00149613 | |
147.02 (110.99 Anhydrous) | |
Hygroscopic | |
Calcium chloride dihydrate, For ACS analysis |
Powder | |
CaCl2·2H2O | |
Soluble in water,acetic acid and alcohol. | |
ACS Reagent | |
0.835 g/mL |
Safety and Handling
P264b-P280i-P305+P351+P338
H319
missing translation for 'einecsNumber' : 233-140-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Calcium chloride dihydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Calcium chloride dihydrate
RUO – Research Use Only