missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride dihydrate, extra pure, flakes for drying purposes, 77-80% CaCl2, Thermo Scientific Chemicals

£69.95 - £152.76

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2·2H2O
Molecular Weight (g/mol) 147.02
MDL Number MFCD00149613
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10657662
View Documents
Thermo Scientific Acros
123350025
2.5 kg Plastic Bottle
£69.95
2.50kg
Please to purchase this item. Need a web account? Register with us today!
10063853
View Documents
Thermo Scientific Acros
123350100
10 kg Plastic drum
£152.76
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
Specifications

Chemical Identifiers

10035-04-8
CaCl2·2H2O
147.02
MFCD00149613

Specifications

175.0°C
Gray-White to White
CaCl2·2H2O
MFCD00149613
15, 1661
147.02
77 to 80% (CaCl2)
0.83
Calcium chloride dihydrate, For drying pursposes
10035-04-8
Flakes
CaCl2·2H2O
01,104
Solubility in water: 1000g/L (0°C). Other solubilities: freely soluble in alcohol
147.02
Extra Pure
0.8300g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • Calcium chloride dihydrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only