missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride dihydrate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99%, Honeywell Fluka™

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99%

£45.04 - £1210.24

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2 · 2H2O
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15683380
View Documents
Honeywell-Fluka
31307-500G
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15663380
View Documents
Honeywell-Fluka
31307-1KG
1 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15673380
View Documents
Honeywell-Fluka
31307-2.5KG
2.5 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15603390
View Documents
Honeywell-Fluka
31307-6X2.5KG
6 x 2.5 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15693380
View Documents
Honeywell-Fluka
31307-50KG
50 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

10035-04-8
CaCl2 · 2H2O

Specifications

10035-04-8
MFCD00149613
147.01g/mol
ACS/puriss. p.a.
CaCl2 · 2H2O
NONH for all modes of transport
≥99%
Calcium Chloride Dihydrate, Reag. Ph. Eur.
Videos
Safety and Handling

Safety and Handling

P305 + P351 + P338

H319

missing translation for 'einecsNumber' : 600-075-5

missing translation for 'rtecsNumber' : EV9810000

Product Identifier
  • Calcium chloride dihydrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications