missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride Dihydrate (White Crystals to Powder), Fisher BioReagents™
Click to view available options
Quantity:
100 g
250 g
500 g
Packaging:
Poly Bottle
Description
Calcium Chloride Dihydrate (White Crystals to Powder)
Calcium Chloride is suitable for use in the preparation of buffer solutions.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 175°C |
Color | Beige-White |
pH | 4.5 to 6.5 |
Physical Form | Solid |
Quantity | 500 g |
Assay Percent Range | 99.0 to 105.0 % |
Merck Index | 15, 1661 |
Formula Weight | 147.02 |
Percent Purity | 99.0 to 105.0% |
Identification | Pass Test |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Calcium Chloride Dihydrate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Calcium chloride, dihydrate, Calcium chloride
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'