missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride EU Pharmacopoeia 0.01M, Fisher Chemical™

Catalog Number 12950384
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Concentration 0.01M
CAS 10043-52-4
Packaging Bottle
Quantity 1000 mL
Molecular Formula CaCl2
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Ca++]
IUPAC Name calcium dichloride
Molecular Weight (g/mol) 110.98
PubChem CID 5284359
ChEBI CHEBI:3312
Grade EU Ph Reference: 1014602
Chemical Name or Material Calcium chloride solution
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'