missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride EU Pharmacopoeia 0.02M, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Specifications
Concentration | 02.M |
CAS | 10035-04-8 |
Packaging | Bottle |
Quantity | 1000 mL |
Type | Mass Spectrometry Standards |
Grade | EU Ph Ref: 1014603 |
Solvent or Matrix | HNO3 |
Chemical Name or Material | Calcium chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'