missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride, Extra Pure, SLR, Fused, Granular, Fisher Chemical™
36.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Description
Calcium Chloride, Extra Pure, SLR, Fused, Granular
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 1600°C |
Melting Point | 782°C |
Color | Beige |
pH | 8 to 10 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 110.99g/mol |
Packaging | Plastic powder jar |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Calcium chloride
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'