missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride hexahydrate, 97+%, extra pure

Product Code. 10610414 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
This item is not returnable. View return policy

Product Code. 10610414

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7774-34-7 | CaCl2·6H2O | 219.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7774-34-7
Molecular Formula CaCl2·6H2O
Molecular Weight (g/mol) 219.08
MDL Number MFCD00149616
InChI Key QHFQAJHNDKBRBO-UHFFFAOYSA-L
Synonym calcium chloride hexahydrate, antarcticite, acmc-20alis, calcium hexahydrate dichloride, calcium chloride-water 1/6, calcium chloride hydrate, puratronic, calcium chloride, hydrated, calcium chloride hydrate trace metals basis 1g
PubChem CID 6093252
IUPAC Name calcium;dichloride;hexahydrate
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2]

Specifications

Melting Point 30.0°C
Color Colorless
Physical Form Chunks
Quantity 1 kg
Assay Percent Range 97.0 to 103% (Complexometry)
Linear Formula CaCl2·6H2O
Merck Index 15, 1661
Solubility Information Solubility in water: 5360g/L (20°C)
Formula Weight 219.08
Percent Purity ≥97%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Calcium chloride hexahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Calcium chloride hexahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.