missing translation for 'onlineSavingsMsg'
Learn More
Calcium Chloride Hexahydrate, 98.5%+, Extra Pure, SLR, Fisher Chemical™ Product Code.: 12648625

Calcium Chloride Hexahydrate, 98.5%+, Extra Pure, SLR, Fisher Chemical™

Product Code. 12648625
1 kg
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 12648625

Brand: Fisher Chemical C/1250/60

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7774-34-7
Molecular Formula CaCl2H12O6
Molecular Weight (g/mol) 219.07
MDL Number 149616
InChI Key QHFQAJHNDKBRBO-UHFFFAOYSA-L
IUPAC Name calcium hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ca++]

Specifications

Melting Point 30°C
Color Colorless
pH 6.5 to 8.0
Physical Form Solid
Quantity 1 kg
Formula Weight 219.08g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Calcium chloride hexahydrate, 98.5%+, Specified (SLR)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium Chloride Hexahydrate, 98.5%+, Extra Pure, SLR, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.