missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride Hexahydrate, 98.5%+, Extra Pure, SLR, Fisher Chemical™

Product Code. p-7072897 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 12648625

Brand: Fisher Chemical C/1250/60

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7774-34-7
Molecular Formula CaCl2H12O6
Molecular Weight (g/mol) 219.07
MDL Number 149616
InChI Key QHFQAJHNDKBRBO-UHFFFAOYSA-L
IUPAC Name calcium hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ca++]

Specifications

Melting Point 30°C
Color Colorless
pH 6.5 to 8.0
Physical Form Solid
Quantity 1 kg
Formula Weight 219.08g/mol
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.