Learn More
Calcium chloride hydrate, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals
£34.87 - £1221.08
Chemical Identifiers
CAS | 22691-02-7 |
---|---|
Molecular Formula | CaCl2H12O6 |
Molecular Weight (g/mol) | 219.068 |
MDL Number | MFCD00149616 |
InChI Key | QHFQAJHNDKBRBO-UHFFFAOYSA-L |
Synonym | calcium chloride hexahydrate, antarcticite, acmc-20alis, calcium hexahydrate dichloride, calcium chloride-water 1/6, calcium chloride hydrate, puratronic, calcium chloride, hydrated, calcium chloride hydrate trace metals basis 1g |
PubChem CID | 6093252 |
IUPAC Name | calcium;dichloride;hexahydrate |
SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11360919
|
Thermo Scientific Alfa Aesar
010680.04 |
2 g |
£34.87
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11330919
|
Thermo Scientific Alfa Aesar
010680.09 |
10 g |
£83.52
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11350919
|
Thermo Scientific Alfa Aesar
010680.18 |
50 g |
£307.53
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11340919
|
Thermo Scientific Alfa Aesar
010680.30 |
250 g |
£1221.08
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a food additive, as an electrolyte, it is used as a buffer in pools, an additive in plastics and in fire extinguishers. It is used as a fabric softener, dust control on roads. Calcium chloride is a soluble compound that can serve as a source of calcium ions in a solution. Calcium plays important roles in many biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
22691-02-7 | |
219.068 | |
QHFQAJHNDKBRBO-UHFFFAOYSA-L | |
6093252 | |
O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2] |
CaCl2H12O6 | |
MFCD00149616 | |
calcium chloride hexahydrate, antarcticite, acmc-20alis, calcium hexahydrate dichloride, calcium chloride-water 1/6, calcium chloride hydrate, puratronic, calcium chloride, hydrated, calcium chloride hydrate trace metals basis 1g | |
calcium;dichloride;hexahydrate |
Specifications
22691-02-7 | |
CaCl2H12O6 | |
14,1659 | |
Soluble in water (5360mg/ml at 20°C),and alcohol. | |
O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2] | |
219.068 | |
110.99 (Anhydrous) | |
Puratronic™ | |
Hygroscopic | |
(metals basis) |
Crystalline Aggregates | |
MFCD00149616 | |
calcium chloride hexahydrate, antarcticite, acmc-20alis, calcium hexahydrate dichloride, calcium chloride-water 1/6, calcium chloride hydrate, puratronic, calcium chloride, hydrated, calcium chloride hydrate trace metals basis 1g | |
QHFQAJHNDKBRBO-UHFFFAOYSA-L | |
calcium;dichloride;hexahydrate | |
6093252 | |
99.9965% | |
Odorless | |
1.71 g/mL | |
Calcium chloride hydrate |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
missing translation for 'einecsNumber' : 607-129-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only