missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride, purum, Honeywell Fluka™

Purum, drying agent, dehydrated, granulated,≥97.0% (KT)

£61.07 - £218.80

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcalShow MoreShow Less
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium;dichloride
SMILES [Cl-].[Cl-].[Ca++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15621930
SDS View Documents
Honeywell-Fluka
21074-1KG
1 kg N/A N/A N/A
15631930
SDS View Documents
Honeywell-Fluka
21074-5KG
5 kg N/A N/A N/A
Description

Description

  • ≥95% for general laboratory use.
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium Chloride, purum, Honeywell Fluka™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.