missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride solution, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Specifications
Concentration | 0.02M (0.04N) |
CAS | 10035-04-8 |
Packaging | Bottle |
Quantity | 1000 mL |
Chemical Name or Material | Calcium Chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'