missing translation for 'onlineSavingsMsg'
Learn More
Calcium Chloride Solution, Honeywell™ Product Code.: 15641930

Calcium Chloride Solution, Honeywell™

Product Code. 15641930 Shop All Honeywell Fluka Products
1 L
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy

Product Code. 15641930

Brand: Honeywell-Fluka 211141L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Volumetric, 1.0 M CaCl2

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Specifications

CAS 10043-52-4
Quantity 1 L
Molecular Formula CaCl2
MDL Number MFCD00010903
UN Number NONH for all modes of transport
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Ca++]
IUPAC Name calcium;dichloride
Molecular Weight (g/mol) 110.98
PubChem CID 5284359
ChEBI CHEBI:3312
Formula Weight 110.98g/mol
Chemical Name or Material Calcium Chloride Solution
Show More Show Less
compliance-icons
Product Identifier
  • Calcium chloride solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : calcium chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium Chloride Solution, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.