missing translation for 'onlineSavingsMsg'
Learn More

Calcium crystalline dendritic pieces, 99.98% (metals basis), Thermo Scientific Chemicals

£199.00 - £798.98

Chemical Identifiers

CAS 7440-70-2
Molecular Formula Ca
Molecular Weight (g/mol) 40.08
MDL Number MFCD00010897 MFCD00085314
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
Synonym unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
PubChem CID 5460341
ChEBI CHEBI:29320
IUPAC Name calcium
SMILES [Ca]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11382408
View Documents
Thermo Scientific Alfa Aesar
014457.06
5 g
£199.00
5g
Please to purchase this item. Need a web account? Register with us today!
11392408
View Documents
Thermo Scientific Alfa Aesar
014457.14
25 g
£798.98
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

7440-70-2
40.08
OYPRJOBELJOOCE-UHFFFAOYSA-N
5460341
calcium
Ca
MFCD00010897 MFCD00085314
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
CHEBI:29320
[Ca]

Specifications

7440-70-2
MFCD00010897 MFCD00085314
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
[Ca]
40.08
CHEBI:29320
Odorless
Pieces
Calcium crystalline dendritic pieces
Ca
UN1401
OYPRJOBELJOOCE-UHFFFAOYSA-N
calcium
5460341
99.98%
Ampule
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H261
In contact with water releases flammable gas.

P223-P231+P232-P280-P335+P334-P370+P378q-P501c

H261

missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: CALCIUM

missing translation for 'einecsNumber' : 213-179-5

missing translation for 'rtecsNumber' : EV8040000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only