missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium dihydrogen phosphate hydrate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Description
Calcium dihydrogen phosphate hydrate is a pH control reagent, stabilizer for plastics, manufacture of glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Calcium dihydrogen phosphate hydrate is a pH control reagent, stabilizer for plastics, manufacture of glass.
Solubility
Soluble in water 18 g/L @ 30°C.
Notes
Loses H{2}O at 100°, dec. at 200°
Calcium dihydrogen phosphate hydrate is a pH control reagent, stabilizer for plastics, manufacture of glass.
Solubility
Soluble in water 18 g/L @ 30°C.
Notes
Loses H{2}O at 100°, dec. at 200°
Specifications
Specifications
Melting Point | ∼200°C (decomposition) |
Quantity | 500 g |
Flash Point | None |
Merck Index | 14,1693 |
Solubility Information | Soluble in water 18g/L @ 30°C. |
Formula Weight | 234.06 (Anhydrous) |
Percent Purity | 97% |
Chemical Name or Material | Calcium dihydrogen phosphate hydrate |
Safety and Handling
Product Identifier
- Calcium dihydrogen phosphate hydrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Phosphoric acid, calcium salt
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction