Learn More
Calcium fluoride, 99.5% (metals basis), Thermo Scientific Chemicals
£77.89 - £782.90
Chemical Identifiers
CAS | 7789-75-5 |
---|---|
Molecular Formula | CaF2 |
Molecular Weight (g/mol) | 78.08 |
MDL Number | MFCD00010907 |
InChI Key | WUKWITHWXAAZEY-UHFFFAOYSA-L |
Synonym | calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade |
PubChem CID | 84512 |
ChEBI | CHEBI:35437 |
SMILES | [F-].[F-].[Ca++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11366419
|
Thermo Scientific Alfa Aesar
011055.30 |
250 g |
£77.89
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376419
|
Thermo Scientific Alfa Aesar
011055.A1 |
1 kg |
£166.83
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11386419
|
Thermo Scientific Alfa Aesar
011055.A7 |
5 kg |
£782.90
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium fluoride is the commercially important source of fluoride compounds as well as hydrogen fluoride. It is transparent from ultraviolet to infrared region. It is used in spectroscopy, thermal imaging systems and excimer lasers. It is also used in the manufacturing of optical components including windows, prisms and lenses. In addition, it is also used in aluminum-metallurgy, brake lining manufacturing industries, and in tooth enamel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-75-5 | |
78.08 | |
WUKWITHWXAAZEY-UHFFFAOYSA-L | |
84512 | |
[F-].[F-].[Ca++] |
CaF2 | |
MFCD00010907 | |
calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade | |
CHEBI:35437 |
Specifications
ca 2500°C | |
7789-75-5 | |
Powder | |
CaF2 | |
EM>5, | |
Slightly soluble in acid. Insoluble in acetone. | |
[F-].[F-].[Ca++] | |
78.08 | |
CHEBI:35437 | |
Odorless | |
Hygroscopic | |
3.18 g/cm3 |
1403°C | |
White | |
99.5% (Metal basis) | |
MFCD00010907 | |
calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade | |
WUKWITHWXAAZEY-UHFFFAOYSA-L | |
calcium;difluoride | |
84512 | |
78.08 | |
1.434 | |
White powder | |
Calcium fluoride |
Safety and Handling
H315-H319-H335-H303
missing translation for 'einecsNumber' : 232-188-7
missing translation for 'rtecsNumber' : EW1760000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only