missing translation for 'onlineSavingsMsg'
Learn More

Calcium fluoride, 99%, pure, anhydrous

Product Code. 13438468 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
10g
500g
This item is not returnable. View return policy

Product Code. 13438468

Brand: Thermo Scientific Acros 218255000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 7789-75-5 | CaF2 | 78.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-75-5
Molecular Formula CaF2
Molecular Weight (g/mol) 78.08
MDL Number MFCD00010907
InChI Key WUKWITHWXAAZEY-UHFFFAOYSA-L
Synonym calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
PubChem CID 84512
ChEBI CHEBI:35437
SMILES [F-].[F-].[Ca++]

Specifications

Boiling Point 2500.0°C
Melting Point 1402.0°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 98.5% min. (CaF2)
Linear Formula CaF2
Fieser 15,287
Merck Index 15,1669
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ammonium salts
IUPAC Name calcium difluoride
Formula Weight 78.08
Percent Purity 99%
Grade Pure
Packaging Plastic bottle
Specific Gravity 3.18
Density 3.1800g/mL
Chemical Name or Material Calcium fluoride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Calcium fluoride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.