Learn More
Calcium fluoride, 99%, Thermo Scientific Chemicals
Description
Calcium fluoride is the commercially important source for fluoride compounds as well as hydrogen fluoride. It is transparent from ultraviolet to infrared region. It is used in spectroscopy, thermal imaging systems and excimer lasers; it is also used in the manufacturing of optical components including windows, prisms and lenses. In addition, it is also used in aluminum-metallurgy, brake lining manufacturing industries, and in tooth enamel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | ≈2500°C |
Melting Point | 1403°C |
Physical Form | Crystals |
Quantity | 500 g |
Beilstein | EM>5, |
Solubility Information | Slightly soluble in acid. Insoluble in acetone. |
IUPAC Name | calcium difluoride |
Formula Weight | 78.08 |
Percent Purity | 99% |
Packaging | Plastic bottle |
Show More |
RUO – Research Use Only