missing translation for 'onlineSavingsMsg'
Learn More

Calcium Formate, 98%, Pure

Product Code. 10679104 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
250g
This item is not returnable. View return policy

Product Code. 10679104

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 544-17-2 | C2H2CaO4 | 130.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-17-2
Molecular Formula C2H2CaO4
Molecular Weight (g/mol) 130.11
MDL Number MFCD00036108
InChI Key CBOCVOKPQGJKKJ-UHFFFAOYSA-L
Synonym calcium formate, calcium diformate, formic acid, calcium salt, calcoform, unii-np3jd65npy, mravencan vapenaty czech, calcium formate ca hco2 2, formic acid calcium salt, np3jd65npy, formic acid, calcium salt 2:1
PubChem CID 10997
ChEBI CHEBI:81851
IUPAC Name calcium;diformate
SMILES C(=O)[O-].C(=O)[O-].[Ca+2]

Specifications

Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (HCOO)2Ca
Quantity 250 g
Merck Index 15,167
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol
Formula Weight 130.11
Percent Purity 98%
Grade Pure
Physical Form Fine Crystalline Powder
Chemical Name or Material Calcium formate
Show More Show Less
compliance-icons
Product Identifier
  • Calcium formate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Calcium formate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.