missing translation for 'onlineSavingsMsg'
Learn More
Calcium granules, -9 mesh, 98.8% (metals basis), Thermo Scientific Chemicals Product Code.: 11328798

Calcium granules, -9 mesh, 98.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11328798
250 g
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11328798

Brand: Thermo Scientific Alfa Aesar 041653.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-70-2
Molecular Formula Ca
Molecular Weight (g/mol) 40.08
MDL Number MFCD00010897 MFCD00085314
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
Synonym unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
PubChem CID 5460341
ChEBI CHEBI:29320
IUPAC Name calcium
SMILES [Ca]

Specifications

Color Silver-White
Appearance Silvery-white granules
Assay Percent Range 98.8% (Metals basis)
Quantity 250 g
Mesh Size -9 Mesh
Odor Odorless
Vapor Pressure 0 hPa
Form Granular
Chemical Name or Material Calcium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX08K007 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number041653.18,041653.A1,041653.30
Lot NumberX08K009 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number041653.18,041653.A1,041653.30
Lot NumberX08K008 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number041653.18,041653.A1,041653.30
Lot NumberX08K005 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number041653.18,041653.A1,041653.30
Lot NumberU26K032 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog Number041653.18,041653.A1,041653.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium granules, -9 mesh, 98.8% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.