missing translation for 'onlineSavingsMsg'
Learn More
Calcium hydride, ca. 93%, extra pure, 0-2 mm grain size, Thermo Scientific Chemicals Product Code.: 10491431

Calcium hydride, ca. 93%, extra pure, 0-2 mm grain size, Thermo Scientific Chemicals

Product Code. 10491431
100 g
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy

Product Code. 10491431

Brand: Thermo Scientific Acros 199901000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-78-8
Molecular Formula CaH2
Molecular Weight (g/mol) 42.09
MDL Number MFCD00010897

Specifications

Melting Point 675.0°C
Quantity 100 g
Assay Percent Range 92% min. (Total Ca)
Linear Formula CaH2
Fieser 01,105
Merck Index 15,1674
Solubility Information Solubility in water: reacts. Other solubilities: decomposes with lower alcohols and carboxylic acid, soluble only in salt melts
Formula Weight 42.09
Percent Purity ∼93%
Grade Extra Pure
Packaging Plastic bottle
Specific Gravity 1.9
Density 1.9000g/mL
Chemical Name or Material Calcium hydride, extra pure, 0-2 mm grain size
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467192 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number199905000,199900025,199901000
Lot NumberA0461915 Certificate TypeCertificate of Analysis Date16/05/2024 Catalog Number199905000,199900025,199901000
Lot NumberA0458367 Certificate TypeCertificate of Analysis Date13/12/2023 Catalog Number199905000,199900025,199901000
Lot NumberA0438173 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number199905000,199900025,199901000
Lot NumberA0396681 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number199905000,199900025,199901000
compliance-icons
Product Identifier
  • Calcium hydride
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
  • P370+P378-In case of fire: Use to extinguish.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium hydride, ca. 93%, extra pure, 0-2 mm grain size, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.