missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydrogen phosphate, anhydrous, 98% min

Product Code. 11389387
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 11389387

Brand: Thermo Scientific Alfa Aesar 040232.A7

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 7757-93-9 | CaHO4P | 136.06 g/mol

It is used as a feed additive for livestock and poultry. It can promote feed digestion, put on animal’s weight, raise laying rate or producing milk rate, cure diseases such as rickets, osteomalacia and anemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-93-9
Molecular Formula CaHO4P
Molecular Weight (g/mol) 136.06
MDL Number MFCD00010909
InChI Key FUFJGUQYACFECW-UHFFFAOYSA-L
Synonym calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1
PubChem CID 24441
ChEBI CHEBI:32596
IUPAC Name calcium;hydrogen phosphate
SMILES [Ca++].OP([O-])([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 5 kg
Linear Formula CaHPO4
Merck Index 14,1692
Solubility Information Soluble in hydrochloric acid,nitric acid,acetic acid,ammonium citrate solution. Slightly soluble in water. Insoluble in ethanol.
Formula Weight 136.06
Percent Purity ≤98%
Odor Odorless
Chemical Name or Material Calcium hydrogen phosphate, Anhydrous

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.