Learn More
Calcium hydroxide, 95%, Thermo Scientific Chemicals
£14.77 - £93.67
Chemical Identifiers
CAS | 1305-62-0 |
---|---|
Molecular Formula | CaH2O2 |
Molecular Weight (g/mol) | 74.09 |
MDL Number | MFCD00010901 |
InChI Key | AXCZMVOFGPJBDE-UHFFFAOYSA-L |
Synonym | calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid |
PubChem CID | 6093208 |
ChEBI | CHEBI:31341 |
IUPAC Name | calcium;dihydroxide |
SMILES | [OH-].[OH-].[Ca++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11460503
|
Thermo Scientific Alfa Aesar
A12650.30 |
250 g |
£14.77
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450503
|
Thermo Scientific Alfa Aesar
A12650.0B |
1000 g |
£23.52
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470503
|
Thermo Scientific Alfa Aesar
A12650.0I |
5000 g |
£93.67
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium hydroxide is used as a flocculant in water and sewage treatment. It is also used in fresh water treatment to raise the pH of the water thereby controlling the corrosion of pipes. It is widely used in the production of calcium stearate. It is an active ingredient in whitewash, mortar and plaster. This compound is also used in the petroleum refining industry for the preparation of additives to oils and in the preparation of dry mixes for painting and decorating. It finds application as an antifungal and antimicrobial preservative for vegetables in storage.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCalcium hydroxide is used as a flocculant in water and sewage treatment. It is also used in fresh water treatment to raise the pH of the water thereby controlling the corrosion of pipes. It is widely used in the production of calcium stearate. It is an active ingredient in whitewash, mortar and plaster. This compound is also used in the petroleum refining industry for the preparation of additives to oils and in the preparation of dry mixes for painting and decorating. It finds application as an antifungal and antimicrobial preservative for vegetables in storage.
Solubility
Soluble in glycerol and acids. Slightly soluble in water.Insoluble in alcohol.
Notes
Air sensitive. Incompatible with strong acids.Incompatible with maleic anhydride, phosphorous, nitroethane, nitromethane, nitorparaffins, nitropropane and strong acids.
Chemical Identifiers
1305-62-0 | |
74.09 | |
AXCZMVOFGPJBDE-UHFFFAOYSA-L | |
6093208 | |
calcium;dihydroxide |
CaH2O2 | |
MFCD00010901 | |
calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid | |
CHEBI:31341 | |
[OH-].[OH-].[Ca++] |
Specifications
1305-62-0 | |
Ca(OH)2 | |
UN3262 | |
calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid | |
AXCZMVOFGPJBDE-UHFFFAOYSA-L | |
calcium;dihydroxide | |
6093208 | |
74.09 | |
Odorless | |
2.24 |
CaH2O2 | |
MFCD00010901 | |
14,1673 | |
Soluble in glycerol and acids. Slightly soluble in water.Insoluble in alcohol. | |
[OH-].[OH-].[Ca++] | |
74.09 | |
CHEBI:31341 | |
95% | |
Air Sensitive | |
Calcium hydroxide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 215-137-3
missing translation for 'rtecsNumber' : EW2800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
- Calcium hydroxide
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : calcium hydroxide
RUO – Research Use Only