missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Hydroxide, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
1 kg
Description
Calcium Hydroxide, Certified AR for Analysis
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 2850°C |
Melting Point | 580°C |
Color | White |
pH | 12.4 |
Physical Form | Solid |
Quantity | 250 g |
Formula Weight | 74.09g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'