missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide, Certified AR for Analysis, Fisher Chemical™

£160.00 - £450.00

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
IUPAC Name calcium dihydroxide
SMILES [OH-].[OH-].[Ca++]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10090070
SDS View Documents
Fisher Chemical
C/1642/50
250 g
£160.00
250g
Please to purchase this item. Need a web account? Register with us today!
10498780
SDS View Documents
Fisher Chemical
C/1642/53
500 g
£244.00
500g
Please to purchase this item. Need a web account? Register with us today!
10283370
SDS View Documents
Fisher Chemical
C/1642/60
1 kg
£450.00
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Calcium Hydroxide, Certified AR for Analysis

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium Hydroxide, Certified AR for Analysis, Fisher Chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.