missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Hydroxide, Reag. Ph. Eur., puriss. p.a., Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
1 kg
25 kg
6 x 1 kg
Unit Size:
100g
1kg
25kg
500g
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 500 g |
| Linear Formula | Ca(OH)2 |
| UN Number | NONH for all modes of transport |
| Formula Weight | 74.09g/mol |
| Percent Purity | ≥96% |
| Grade | puriss. p.a. |
| Density | 2.24g/mL at 25°C |
| Chemical Name or Material | Calcium Hydroxide, Reag. Ph. Eur. |
Safety and Handling
Product Identifier
- Calcium hydroxide
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Calcium dihydroxide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction