missing translation for 'onlineSavingsMsg'
Learn More

Calcium hypochlorite, tech., Thermo Scientific Chemicals

Product Code. 11319148
Click to view available options
Quantity:
10 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

For Use in Deodorant, Oxidizing Agent, Bleaching Agent

Deodorant, oxidizing agent, bleaching agentCalcium hypochlorite is used for swimming pool sanitation and other water treatment plants. It is also used to disinfect drinking water, equipments in the industry and surfaces and equipments in kitchens. Further, it is used to bleach textiles and paper. It is also employed in bathroom cleaners and laundry detergents. It is used as an oxidizing agent in organic chemistry. It is involved in the cleavage of glycols, alfa-hydroxy carboxylic acids and keto acids to yield fragmented aldehydes or carboxylic acids respectively. It plays an important role in the haloform reaction to prepare chloroform.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-54-3
Molecular Formula CaCl2O2
MDL Number MFCD00010900
InChI Key ZKQDCIXGCQPQNV-UHFFFAOYSA-N
Synonym calcium hypochlorite, calcium hypochloride, bleaching powder, chlorinated lime, hypochlorous acid, calcium salt, caporit, losantin, pittchlor, pittcide, pittclor
PubChem CID 24504

Specifications

Color White
Physical Form Granules
Quantity 10 g
SMILES [O-]Cl.[O-]Cl.[Ca+2]
IUPAC Name calcium;dihypochlorite
Molecular Weight (g/mol) 142.976
Formula Weight 142.98
Appearance White granular powder
Density 2.35g/cm3 at 20°C
Chemical Name or Material Calcium hypochlorite

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'