missing translation for 'onlineSavingsMsg'
Learn More
Calcium ingot, 97% (metals basis), Thermo Scientific Chemicals Product Code.: 11381178

Calcium ingot, 97% (metals basis), Thermo Scientific Chemicals

Product Code. 11381178
700 g
Click to view available options
Quantity:
700 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11381178

Brand: Thermo Scientific Alfa Aesar 010128.39

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-70-2
Density 1.54
Assay Percent Range 99% (metals basis)
Molecular Formula Ca
MDL Number MFCD00010897 MFCD00085314
Quantity 700 g
Synonym unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
SMILES [Ca]
IUPAC Name calcium
Molecular Weight (g/mol) 40.08
PubChem CID 5460341
ChEBI CHEBI:29320
Formula Weight 40.078
Form Ingot
Chemical Name or Material Calcium
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN19K040 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog Number010128.8Z,010128.39
Lot NumberT12C013 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010128.8Z,010128.39
Lot NumberE05K18 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010128.8Z,010128.39
Lot NumberK21Q052 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010128.8Z,010128.39
Lot NumberF07J26 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010128.8Z,010128.39

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium ingot, 97% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.