missing translation for 'onlineSavingsMsg'
Learn More

Calcium iodate, pure, Thermo Scientific Chemicals

Oxidation reagent

£75.48 - £283.41

Chemical Identifiers

CAS 7789-80-2
Molecular Formula CaI2O6
Molecular Weight (g/mol) 389.89
InChI Key UHWJJLGTKIWIJO-UHFFFAOYSA-L
Synonym calcium iodate, lautarite, calcium diiodate, unii-l8mn4y57br, l8mn4y57br, iodic acid hio3 , calcium salt 2:1, autarite, iodic acid, calcium salt, calcium iodate ca io3 2, hsdb 986
PubChem CID 24619
IUPAC Name calcium;diiodate
SMILES [O-]I(=O)=O.[O-]I(=O)=O.[Ca+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15570534
SDS View Documents
Thermo Scientific Acros
318415000
500 g
£75.48
500g
Please to purchase this item. Need a web account? Register with us today!
15560534
SDS View Documents
Thermo Scientific Acros
318410025
2.5 kg
£283.41
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium iodate, pure, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.