missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium L-threonate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
250 g
500 g
Description
Calcium L-threonate is reactant involved in synthesis of cyclic hydroxamates and substituted oxazinanones and oxidative degradation to yield oxalate complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Calcium L-threonate is reactant involved in synthesis of cyclic hydroxamates and substituted oxazinanones and oxidative degradation to yield oxalate complexes.
Solubility
Soluble in water. Sparingly soluble in methanol.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Calcium L-threonate is reactant involved in synthesis of cyclic hydroxamates and substituted oxazinanones and oxidative degradation to yield oxalate complexes.
Solubility
Soluble in water. Sparingly soluble in methanol.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | >300°C |
Color | Brown |
Quantity | 500 g |
Solubility Information | Soluble in water. Sparingly soluble in methanol. |
Formula Weight | 310.27 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | Calcium L-threonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'