missing translation for 'onlineSavingsMsg'
Learn More

Calcium Lactate Pentahydrate, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™

£98.80 - £481.00

Chemical Identifiers

CAS 5743-47-5
Molecular Formula C6H20CaO11
Molecular Weight (g/mol) 308.29
MDL Number 150987
InChI Key HPVJXNNKHRNBOY-UHFFFAOYNA-L
IUPAC Name calcium bis(2-hydroxypropanoate) pentahydrate
SMILES O.O.O.O.O.[Ca++].CC(O)C([O-])=O.CC(O)C([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10385310
View Documents
Fisher Chemical
C/1820/53
500 g Plastic powder jar This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12626677
View Documents
Fisher Chemical
C/1820/65
5 kg Plastic keg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

5743-47-5
308.29
HPVJXNNKHRNBOY-UHFFFAOYNA-L
O.O.O.O.O.[Ca++].CC(O)C([O-])=O.CC(O)C([O-])=O
C6H20CaO11
150987
calcium bis(2-hydroxypropanoate) pentahydrate

Specifications

240°C
Solid
150987
O.O.O.O.O.[Ca++].CC(O)C([O-])=O.CC(O)C([O-])=O
308.29
5743-47-5
C6H20CaO11
HPVJXNNKHRNBOY-UHFFFAOYNA-L
calcium bis(2-hydroxypropanoate) pentahydrate
308.29g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Calcium lactate pentahydrate

SDS
Documents

Documents

Product Certifications