Learn More
Calcium molybdenum oxide, 99.8%, Thermo Scientific™
£31.00 - £83.70
Chemical Identifiers
CAS | 7789-82-4 |
---|---|
Molecular Formula | CaMoO4 |
Molecular Weight (g/mol) | 200.02 |
MDL Number | MFCD00015976 |
InChI Key | BIOOACNPATUQFW-UHFFFAOYSA-N |
Synonym | calcium molybdate, molybdate, calcium, calcium molybdate vi, unii-q03c83v2an, calcium molybdenum oxide, molybdic acid h2moo4 , calcium salt 1:1, molybdate moo42-, calcium 1:1 , t-4, molybdate moo42-, calcium 1:1 , beta-4, calcium dioxido dioxo molybdenum, calcium 2+ molybdate |
PubChem CID | 24620 |
IUPAC Name | calcium;dioxido(dioxo)molybdenum |
SMILES | [Ca++].[O-][Mo]([O-])(=O)=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7789-82-4 | |
200.02 | |
BIOOACNPATUQFW-UHFFFAOYSA-N | |
24620 | |
[Ca++].[O-][Mo]([O-])(=O)=O |
CaMoO4 | |
MFCD00015976 | |
calcium molybdate, molybdate, calcium, calcium molybdate vi, unii-q03c83v2an, calcium molybdenum oxide, molybdic acid h2moo4 , calcium salt 1:1, molybdate moo42-, calcium 1:1 , t-4, molybdate moo42-, calcium 1:1 , beta-4, calcium dioxido dioxo molybdenum, calcium 2+ molybdate | |
calcium;dioxido(dioxo)molybdenum |
Specifications
965°C | |
White | |
CaMoO4 | |
14,1681 | |
Soluble in mineral acids. Insoluble in water,ethanol,ethyl ether. | |
[Ca++].[O-][Mo]([O-])(=O)=O | |
200.02 | |
200.01 | |
Odorless | |
Calcium molybdenum oxide |
7789-82-4 | |
Powder | |
MFCD00015976 | |
calcium molybdate, molybdate, calcium, calcium molybdate vi, unii-q03c83v2an, calcium molybdenum oxide, molybdic acid h2moo4 , calcium salt 1:1, molybdate moo42-, calcium 1:1 , t-4, molybdate moo42-, calcium 1:1 , beta-4, calcium dioxido dioxo molybdenum, calcium 2+ molybdate | |
BIOOACNPATUQFW-UHFFFAOYSA-N | |
calcium;dioxido(dioxo)molybdenum | |
24620 | |
99.8% | |
4.45 g/mL |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 232-192-9
missing translation for 'rtecsNumber' : EW2975000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures