Learn More
Calcium nitrate tetrahydrate, 97+%, Thermo Scientific Chemicals
£22.19 - £97.47
Chemical Identifiers
CAS | 13477-34-4 |
---|---|
Molecular Formula | CaH8N2O10 |
Molecular Weight (g/mol) | 236.15 |
MDL Number | MFCD00149604 |
InChI Key | MWGCGFYACDTFSB-UHFFFAOYSA-N |
Synonym | calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate |
PubChem CID | 16211656 |
ChEBI | CHEBI:86159 |
IUPAC Name | calcium bis(nitric acid) tetrahydrate |
SMILES | O.[Ca++].[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11354569
|
Thermo Scientific Alfa Aesar
A16645.22 |
100 g |
£22.19
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11457977
|
Thermo Scientific Alfa Aesar
A16645.36 |
500 g |
£29.58
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11467977
|
Thermo Scientific Alfa Aesar
A16645.0E |
2500 g |
£97.47
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium nitrate tetrahydrate is used as a nitrogenous fertilizer and a corrosive inhibitor in diesel fuels. It is utilized in plant cells for culture experiments. It is also used in cooling baths and waste water handling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCalcium nitrate tetrahydrate is used as a nitrogenous fertilizer and a corrosive inhibitor in diesel fuels. It is utilized in plant cells for culture experiments. It is also used in cooling baths and waste water handling.
Solubility
Soluble in water, alcohol and acetone.
Notes
Store in cool place. Hygroscopic. Incompatible with strong reducing agents.
Identifiants chimiques
13477-34-4 | |
236.15 | |
MWGCGFYACDTFSB-UHFFFAOYSA-N | |
16211656 | |
calcium bis(nitric acid) tetrahydrate |
CaH8N2O10 | |
MFCD00149604 | |
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
CHEBI:86159 | |
O.[Ca++].[O-][N+]([O-])=O |
Spécification
132°C (decomposition) | |
13477-34-4 | |
MFCD00149604 | |
14,1682 | |
Soluble in water,alcohol and acetone. | |
O.[Ca++].[O-][N+]([O-])=O | |
236.15 | |
CHEBI:86159 | |
99% | |
Hygroscopic | |
Calcium nitrate tetrahydrate |
43°C to 45°C | |
CaH8N2O10 | |
UN1454 | |
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
MWGCGFYACDTFSB-UHFFFAOYSA-N | |
calcium bis(nitric acid) tetrahydrate | |
16211656 | |
236.15 (164.09 Anhydrous) | |
Odorless | |
1.82 |
Sécurité et traitement
GHS H Statement
H272-H315-H319-H335
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: CALCIUM NITRATE
missing translation for 'einecsNumber' : 233-332-1
missing translation for 'rtecsNumber' : EW3000000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Calcium nitrate, tetrahydrate
Mention d'avertissement
- Danger
Catégorie de danger
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Mention de danger
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Conseils de prudence
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.