Learn More
Calcium nitrate tetrahydrate, 99.98% (metals basis), Thermo Scientific Chemicals
£55.68 - £186.93
Chemical Identifiers
CAS | 13477-34-4 |
---|---|
Molecular Formula | CaH8N2O10 |
Molecular Weight (g/mol) | 236.146 |
MDL Number | MFCD00149604 |
InChI Key | ICSSIKVYVJQJND-UHFFFAOYSA-N |
Synonym | calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronateShow More |
PubChem CID | 16211656 |
ChEBI | CHEBI:86159 |
IUPAC Name | calcium;dinitrate;tetrahydrate |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387278
|
Thermo Scientific Alfa Aesar
030482.14 |
25 g |
£55.68
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397278
|
Thermo Scientific Alfa Aesar
030482.22 |
100 g |
£186.93
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium nitrate tetrahydrate is used as a nitrogenous fertilizer and a corrosive inhibitor in diesel fuels. It is utilized in plant cells for culture experiments. It is also used in cooling baths and waste water handling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13477-34-4 | |
236.146 | |
ICSSIKVYVJQJND-UHFFFAOYSA-N | |
16211656 | |
calcium;dinitrate;tetrahydrate |
CaH8N2O10 | |
MFCD00149604 | |
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronateShow More | |
CHEBI:86159 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2] |
Specifications
132°C (decomposition) | |
13477-34-4 | |
Crystalline | |
CaH8N2O10 | |
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
ICSSIKVYVJQJND-UHFFFAOYSA-N | |
calcium;dinitrate;tetrahydrate | |
16211656 | |
236.15 (164.09 Anhydrous) | |
Hygroscopic | |
1.82 g/cm3 at 20°C |
42°C | |
White | |
99.98% | |
MFCD00149604 | |
Soluble in water,alcohol and acetone. | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2] | |
236.146 | |
CHEBI:86159 | |
Odorless | |
White crystalline or powder | |
Calcium nitrate tetrahydrate |
Safety and Handling
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: III
missing translation for 'einecsNumber' : 233-332-1
missing translation for 'rtecsNumber' : EW3000000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Calcium nitrate tetrahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Calcium nitrate tetrahydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.