missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Nitrate Tetrahydrate, Extra Pure, SLR, Fisher Chemical™
£416.00
Chemical Identifiers
CAS | 13477-34-4 |
---|---|
Molecular Formula | CaH10N2O10 |
Molecular Weight (g/mol) | 238.16 |
MDL Number | 149604 |
InChI Key | DKCJOKYHYIAZQH-UHFFFAOYSA-N |
IUPAC Name | calcium bis(nitric acid) tetrahydrate |
SMILES | O.O.O.O.[Ca++].O[N+]([O-])=O.O[N+]([O-])=O |
Description
Calcium Nitrate Tetrahydrate, Extra Pure, SLR
Chemical Identifiers
13477-34-4 | |
238.16 | |
DKCJOKYHYIAZQH-UHFFFAOYSA-N | |
O.O.O.O.[Ca++].O[N+]([O-])=O.O[N+]([O-])=O |
CaH10N2O10 | |
149604 | |
calcium bis(nitric acid) tetrahydrate |
Specifications
132°C | |
13477-34-4 | |
5 to 7 | |
CaH10N2O10 | |
DKCJOKYHYIAZQH-UHFFFAOYSA-N | |
calcium bis(nitric acid) tetrahydrate | |
236.15g/mol |
45°C | |
White | |
Crystalline Solid | |
149604 | |
O.O.O.O.[Ca++].O[N+]([O-])=O.O[N+]([O-])=O | |
238.16 | |
Plastic powder jar |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Calcium nitrate tetrahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Calcium Nitrate Tetrahydrate, Extra Pure, SLR, Fisher Chemical™