missing translation for 'onlineSavingsMsg'
Learn More

Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals

Product Code. 10537742
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5794-28-5
Molecular Formula C2H2CaO5
Molecular Weight (g/mol) 146.11
MDL Number MFCD00150039
InChI Key LQHWSGSWNOHVHO-UHFFFAOYSA-L
Synonym calcium oxalate monohydrate, unii-4pp86kk527, calcium hydrate oxalate, calcium ethanedioate hydrate 1:1:1, acmc-1bmv0, ethanedioic acid, calcium salt 1:1 , monohydrate, calcium oxalate monohydrate 100g, ethanedioic acid,calcium salt, hydrate 1:?:?
PubChem CID 165350
IUPAC Name calcium;oxalate;hydrate
SMILES O.[Ca++].[O-]C(=O)C([O-])=O

Specifications

Melting Point 200.0°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 98%
Linear Formula (COO)2Ca·H2O
UN Number 2811
Merck Index 14, 1685
Solubility Information Solubility in water: practically insoluble. Other solubilities: practically insoluble in acetic acid,soluble in dilute hcl or hno3
Formula Weight 146.11
Percent Purity 98%
Grade Extra Pure
Loss on Drying 13% max. (200°C)
Packaging Glass bottle
Chemical Name or Material Calcium oxalate monohydrate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Calcium oxalate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'