Learn More
Calcium oxide, 96%, extra pure, Thermo Scientific Chemicals
£25.02 - £51.96
Chemical Identifiers
CAS | 1305-78-8 |
---|---|
Molecular Formula | CaO |
Molecular Weight (g/mol) | 56.08 |
MDL Number | MFCD00010911 |
InChI Key | BRPQOXSCLDDYGP-UHFFFAOYSA-N |
Synonym | calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx |
PubChem CID | 14778 |
ChEBI | CHEBI:31344 |
IUPAC Name | oxocalcium |
SMILES | [O--].[Ca++] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10697532
|
Thermo Scientific Acros
196910010 |
1 kg | Glass bottle |
£25.02
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10542934
|
Thermo Scientific Acros
196910025 |
2.5 kg | Plastic bottle |
£51.96
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
AdsorbentChemical Identifiers
1305-78-8 | |
56.08 | |
BRPQOXSCLDDYGP-UHFFFAOYSA-N | |
14778 | |
oxocalcium |
CaO | |
MFCD00010911 | |
calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx | |
CHEBI:31344 | |
[O--].[Ca++] |
Specifications
2850.0°C | |
1305-78-8 | |
Powder | |
CaO | |
MFCD00010911 | |
calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx | |
BRPQOXSCLDDYGP-UHFFFAOYSA-N | |
oxocalcium | |
14778 | |
56.08 | |
Extra Pure | |
Calcium oxide |
2570.0°C | |
Beige | |
95% min. (Complexometry) | |
CaO | |
15, 1688 | |
Solubility in water: reacts. Other solubilities: soluble in acids (reacts), soluble in glycerol, insoluble in alcohol | |
[O--].[Ca++] | |
56.08 | |
CHEBI:31344 | |
96% | |
3.5% max. |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 215-138-9
- Calcium oxide
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only