missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Oxide, Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
20 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Fine Powder |
Quantity | 1 kg |
Linear Formula | CaO |
UN Number | 1910 |
IUPAC Name | calcium oxidandiide |
Formula Weight | 56.08g/mol |
Percent Purity | 96 to 100.5% (ex ignited substance) |
Grade | puriss. |
Density | 3.3g/mL at 25°C |
Chemical Name or Material | Calcium Oxide, Meets analytical specification of FCC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'