missing translation for 'onlineSavingsMsg'
Learn More

Calcium oxide, Puratronic™, 99.998% (metals basis, excluding other alkaline earth and alkali metals 130ppm max), Thermo Scientific Chemicals

Catalog Number 11350929
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
This item is not returnable. View return policy
1305-78-8
CaO
56.08
MFCD00010911
BRPQOXSCLDDYGP-UHFFFAOYSA-N
calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx
14778
CHEBI:31344
[O--].[Ca++]
This item is not returnable. View return policy
1305-78-8
CaO
56.08
MFCD00010911
BRPQOXSCLDDYGP-UHFFFAOYSA-N
calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx
14778
CHEBI:31344
[O--].[Ca++]

Calcium oxide has wide-ranging utility in industry. It is used in making porcelain, cements, mortars, and glass. It is also employed in flue-gas desulfurization in power plants, purifying sugar, drying the solvents/reagents, preparing bleaching powder, calcium carbide, and calcium cyanamide. It also finds use in water softeners, chemical recovery at Kraft pulp mills, biodiesel production, and in petroleum industry for the detection of traces of water. In agriculture it is used to restore the pH of acidic soils (liming). It is useful as a basic catalyst & acid scavenger in several organic transformations, for example, in Arndt-Eistert Synthesis of alpha-diazoketones, synthesis of heterocycles and oximes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 2850°C
Melting Point 2572°C
Color White
Physical Form Powder
Quantity 2 g
Assay Percent Range 99.998% (Metal basis)
Solubility Information Slightly soluble in water. Insoluble in ethanol. Soluble in glycerol.
IUPAC Name oxocalcium
Formula Weight 56.08
Odor Odorless
Refractive Index 1.83
Sensitivity Air and moisture sensitive
Appearance White powder
Density 3.37
Chemical Name or Material Calcium oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'