Learn More
Calcium perchlorate tetrahydrate, pure, Thermo Scientific Chemicals
£31.86 - £110.55
Chemical Identifiers
CAS | 15627-86-8 |
---|---|
Molecular Formula | CaCl2O8·4H2O |
Molecular Weight (g/mol) | 311.04 |
MDL Number | MFCD00191895 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10055153
|
Thermo Scientific
390631000 |
100 g | Glass bottle |
£31.86
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10310144
|
Thermo Scientific Acros
390635000 |
500 g | Glass bottle |
£110.55
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
15627-86-8 | |
CaCl2O8·4H2O | |
311.04 | |
MFCD00191895 |
Specifications
15627-86-8 | |
Crystalline Powder and/or Crystals | |
CaCl2O8·4H2O | |
Solubility in water: soluble. Other solubilities: soluble in alcohol | |
311.04 | |
Pure |
White | |
CaCl2O8·4H2O | |
MFCD00191895 | |
311.04 | |
12.2 to 13.5% (Calcium) | |
Calcium perchlorate tetrahydrate |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May intensify fire; oxidizer.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Calcium perchlorate tetrahydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only