missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium phosphate, 96%, pure, mixture of Calcium phosphates, Thermo Scientific Chemicals
£57.08 - £208.04
Chemical Identifiers
CAS | 7758-87-4 |
---|---|
Molecular Formula | Ca3O8P2 |
Molecular Weight (g/mol) | 310.18 |
InChI Key | QORWJWZARLRLPR-UHFFFAOYSA-H |
Synonym | calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic |
PubChem CID | 24456 |
ChEBI | CHEBI:9679 |
IUPAC Name | tricalcium;diphosphate |
SMILES | [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12065390
|
Thermo Scientific Acros
446390010 |
1 kg |
£57.08
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12075390
|
Thermo Scientific Acros
446390050 |
5 kg |
£208.04
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7758-87-4 | |
310.18 | |
calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic | |
CHEBI:9679 | |
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2] |
Ca3O8P2 | |
QORWJWZARLRLPR-UHFFFAOYSA-H | |
24456 | |
tricalcium;diphosphate |
Specifications
1670.0°C | |
95% min. (Complexometry) | |
Ca3(PO4)2 | |
calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic | |
QORWJWZARLRLPR-UHFFFAOYSA-H | |
tricalcium;diphosphate | |
24456 | |
310.18 | |
Pure | |
Calcium phosphate |
7758-87-4 | |
Ca3O8P2 | |
15,1696 | |
Solubility in water: 0.1g/L (25°C) | |
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2] | |
310.18 | |
CHEBI:9679 | |
96% | |
Glass bottle |
Safety and Handling
missing translation for 'einecsNumber' : 231-840-8
Product Identifier
- Calcium phosphate
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title