missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium phosphate, for analysis, 35-40% (Ca), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 1670.0°C |
Quantity | 5 kg |
Assay Percent Range | 35-40% |
Linear Formula | Ca3(PO4)2 |
Merck Index | 15,1696 |
Solubility Information | Solubility in water: 0.1g/L (25°C) |
Formula Weight | 310.18 |
Percent Purity | 35 to 40% (Ca) |
Grade | Analytical |
Packaging | Plastic Drum |
Show More |
Safety and Handling
Product Identifier
- Calcium phosphate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'