missing translation for 'onlineSavingsMsg'
Learn More

Calcium phosphate, dibasic, 98+%, for analysis, anhydrous

Product Code. 12665027 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 12665027

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7757-93-9 | CaHO4P | 136.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-93-9
Molecular Formula CaHO4P
Molecular Weight (g/mol) 136.06
MDL Number MFCD00010909
InChI Key FUFJGUQYACFECW-UHFFFAOYSA-L
Synonym calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1
PubChem CID 24441
ChEBI CHEBI:32596
IUPAC Name calcium;hydrogen phosphate
SMILES [Ca++].OP([O-])([O-])=O

Specifications

Quantity 250 g
Assay Percent Range 98.0 to 103% (Complexometry)
Linear Formula CaHPO4
Merck Index 15,1694
Solubility Information Solubility in water: 0.1g/L (25°C). Other solubilities: practically insoluble in alcohol
Formula Weight 136.06
Percent Purity ≥98%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Calcium phosphate, dibasic

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.