missing translation for 'onlineSavingsMsg'
Learn More

Calcium phosphate, dibasic dihydrate, 98+%, extra pure

Product Code. 10549423 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 10549423

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-77-7 | CaHO42H2O | 172.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-77-7
Molecular Formula CaHO42H2O
Molecular Weight (g/mol) 172.09
MDL Number MFCD00149621
InChI Key XAAHAAMILDNBPS-UHFFFAOYSA-L
Synonym calcium hydrogenphosphate dihydrate, calcium hydrogen phosphate dihydrate, unii-o7tsz97gep, calcium monohydrogen phosphate dihydrate, o7tsz97gep, calcium phosphate dibasic dihydrate, dibasic calcium phosphate dihydrate, phosphoric acid, calcium salt 1:1 , dihydrate, brushite, aksepillen
PubChem CID 104805
ChEBI CHEBI:4496
IUPAC Name calcium;hydrogen phosphate;dihydrate
SMILES O.O.OP(=O)([O-])[O-].[Ca+2]

Specifications

Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 98.0 to 105% (on dried substance) (Complexometry)
Linear Formula CaHPO4·2H2O
Solubility Information Solubility in water: 0.2g/L (25°C)
Formula Weight 172.09
Percent Purity ≥98%
Grade Extra Pure
Identification Calcium: positive, Phosphate: positive
Loss on Ignition 24.5% to 26.5% (800°C)
Packaging Plastic bottle
Chemical Name or Material Calcium phosphate, dibasic dihydrate
Show More Show Less
compliance-icons
Product Identifier
  • Calcium phosphate, dibasic dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.