missing translation for 'onlineSavingsMsg'
Learn More
Calcium Phosphate Dibasic, puriss. p.a., >98.5% (KT), Honeywell Fluka™ Product Code.: 15601940

Calcium Phosphate Dibasic, puriss. p.a., >98.5% (KT), Honeywell Fluka™

Product Code. 15601940 Shop All Honeywell Fluka Products
250 g, Poly Bottle
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 15601940

Brand: Honeywell-Fluka 21177250G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., anhydrous, >98.5% (KT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-93-9
Molecular Formula CaHO4P
Molecular Weight (g/mol) 136.06
MDL Number MFCD00010909
InChI Key FUFJGUQYACFECW-UHFFFAOYSA-L
Synonym calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1Show MoreShow Less
PubChem CID 24441
ChEBI CHEBI:32596
IUPAC Name calcium;hydrogen phosphate
SMILES [Ca++].OP([O-])([O-])=O

Specifications

Melting Point ≥450°C
Color White
Physical Form Anhydrous
Quantity 250 g
Linear Formula CaHPO4
UN Number NONH for all modes of transport
Formula Weight 136.06g/mol
Percent Purity >98.5% (KT)
Grade puriss. p.a.
Packaging Poly Bottle
Appearance Powder
Chemical Name or Material Calcium Phosphate Dibasic
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium Phosphate Dibasic, puriss. p.a., >98.5% (KT), Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.