Learn More
Calcium phosphate (pyro), 96% min, Thermo Scientific Chemicals
£21.10 - £158.00
Chemical Identifiers
CAS | 7790-76-3 |
---|---|
Molecular Formula | Ca2O7P2 |
Molecular Weight (g/mol) | 254.10 |
MDL Number | MFCD00015983 |
InChI Key | JUNWLZAGQLJVLR-UHFFFAOYSA-J |
Synonym | calcium pyrophosphate, calcium diphosphate, dicalcium diphosphate, unii-x69nu20d19, dicalcium pyrophosphate, alpha-calcium pyrophosphate, calcium phosphate ca2p2o7, diphosphoric acid, calcium salt, calcium pyrophosphate ca2p2o7, diphosphoric acid, calcium salt 1:2 |
PubChem CID | 24632 |
ChEBI | CHEBI:32598 |
IUPAC Name | dicalcium;phosphonato phosphate |
SMILES | [Ca++].[Ca++].[O-]P([O-])(=O)OP([O-])([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11413840
|
Thermo Scientific Alfa Aesar
089836.30 |
250 g |
£21.10
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493830
|
Thermo Scientific Alfa Aesar
089836.A1 |
1 kg |
£64.20
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11403840
|
Thermo Scientific Alfa Aesar
089836.A7 |
5 kg |
£158.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium phosphate is used in the production of phosphoric acid and fertilizers. It is used in baking as the acid in a leavening agent. It is also used in cheese products. This compound has also been used in gene transfection of cells. It is also being used in the development of contrast agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7790-76-3 | |
254.10 | |
JUNWLZAGQLJVLR-UHFFFAOYSA-J | |
24632 | |
dicalcium;phosphonato phosphate |
Ca2O7P2 | |
MFCD00015983 | |
calcium pyrophosphate, calcium diphosphate, dicalcium diphosphate, unii-x69nu20d19, dicalcium pyrophosphate, alpha-calcium pyrophosphate, calcium phosphate ca2p2o7, diphosphoric acid, calcium salt, calcium pyrophosphate ca2p2o7, diphosphoric acid, calcium salt 1:2 | |
CHEBI:32598 | |
[Ca++].[Ca++].[O-]P([O-])(=O)OP([O-])([O-])=O |
Specifications
1230°C | |
White | |
Ca2O7P2 | |
calcium pyrophosphate, calcium diphosphate, dicalcium diphosphate, unii-x69nu20d19, dicalcium pyrophosphate, alpha-calcium pyrophosphate, calcium phosphate ca2p2o7, diphosphoric acid, calcium salt, calcium pyrophosphate ca2p2o7, diphosphoric acid, calcium salt 1:2 | |
JUNWLZAGQLJVLR-UHFFFAOYSA-J | |
dicalcium;phosphonato phosphate | |
24632 | |
254.1 | |
Odorless | |
Calcium phosphate (pyro) |
7790-76-3 | |
Powder | |
MFCD00015983 | |
Soluble in hydrochloric acid and nitric acid. Insoluble in water. | |
[Ca++].[Ca++].[O-]P([O-])(=O)OP([O-])([O-])=O | |
254.10 | |
CHEBI:32598 | |
≥96% | |
3.09 g/mL |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 232-221-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only