missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium phosphate tribasic, 34-40% Ca, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2 kg
10 kg
Description
Utilized in powdered spices as an anticaking agent and baby powder. It is the starting material for the production of phosphoric acid and fertilizers. Employed in tissue replacement for repairing bony defects and act as drug carrier for drug delivery to the bone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 34-40% Ca |
Solubility Information | Partially soluble in water. |
Formula Weight | 1004.67 |
Appearance | White powder |
Density | 3.14g/cm3 at 20°C |
Chemical Name or Material | Calcium phosphate tribasic, tribasic |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'