Learn More
Chemical Identifiers
CAS | 4075-81-4 |
---|---|
Molecular Formula | C6H10CaO4 |
Molecular Weight (g/mol) | 186.22 |
MDL Number | MFCD00167354 |
InChI Key | BCZXFFBUYPCTSJ-UHFFFAOYSA-L |
Synonym | calcium propionate, calcium dipropionate, calcium propanoate, mycoban, propanoic acid, calcium salt, bioban-c, caswell no. 151, propionic acid, calcium salt, unii-8ai80040kw, hsdb 907 |
PubChem CID | 19999 |
ChEBI | CHEBI:81716 |
IUPAC Name | calcium;propanoate |
SMILES | [Ca++].CCC([O-])=O.CCC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10772202
|
Thermo Scientific Acros
403900010 |
1 kg | Plastic bottle |
£39.70
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10268333
|
Thermo Scientific Acros
403900050 |
5 kg | Plastic drum |
£137.69
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10618904
|
Thermo Scientific Acros
|
25 kg | Fibre drum |
£435.17
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4075-81-4 | |
186.22 | |
BCZXFFBUYPCTSJ-UHFFFAOYSA-L | |
19999 | |
calcium;propanoate |
C6H10CaO4 | |
MFCD00167354 | |
calcium propionate, calcium dipropionate, calcium propanoate, mycoban, propanoic acid, calcium salt, bioban-c, caswell no. 151, propionic acid, calcium salt, unii-8ai80040kw, hsdb 907 | |
CHEBI:81716 | |
[Ca++].CCC([O-])=O.CCC([O-])=O |
Specifications
4075-81-4 | |
White | |
4% max. (105°C, 2 Hrs) | |
98% min. (on dry substance) (Complexometry) | |
(CH3CH2COO)2Ca | |
15, 1700 | |
Solubility in water: 260g/L (20°C). Other solubilities: slightly soluble in methanol and ethanol,practically insoluble in acetone and benzene | |
[Ca++].CCC([O-])=O.CCC([O-])=O | |
186.22 | |
CHEBI:81716 | |
98+% | |
Crystalline Powder or Crystals |
>300.0°C | |
6.9 to 9.0 (10% soln.) | |
Authentic | |
C6H10CaO4 | |
MFCD00167354 | |
calcium propionate, calcium dipropionate, calcium propanoate, mycoban, propanoic acid, calcium salt, bioban-c, caswell no. 151, propionic acid, calcium salt, unii-8ai80040kw, hsdb 907 | |
BCZXFFBUYPCTSJ-UHFFFAOYSA-L | |
calcium;propanoate | |
19999 | |
186.22 | |
0.3% max. (in water) | |
Calcium propionate |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 223-795-8
- Calcium propionate
RUO – Research Use Only