missing translation for 'onlineSavingsMsg'
Learn More

Calcium shot, 99.5% (metals basis), Thermo Scientific™

Product Code. 11361178
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
50 g
250 g
2 x 500 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-70-2
Molecular Formula Ca
Molecular Weight (g/mol) 40.08
MDL Number MFCD00010897 MFCD00085314
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
Synonym unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
PubChem CID 5460341
ChEBI CHEBI:29320
IUPAC Name calcium
SMILES [Ca]

Specifications

Color Silver-White
Appearance Silvery-white shot
Assay Percent Range 99.5% (Metals basis)
Quantity 250 g
Odor Odorless
Vapor Pressure 0 hPa
Form Shot
Chemical Name or Material Calcium
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'